missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Lockout Compliance Manual

Catalog No. 19016497
Change view
Click to view available options
Product Type:
Lockout Binder Vol II
Lockout Compliance Manual
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19016497 Lockout Compliance Manual
189991329 Lockout Binder Vol II
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19016497 Supplier Brady™ Supplier No. 65558

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Lockout Compliance Manual is a binder that keeps your Hazardous Energy Control Procedures organized and protected.

  • RoHS compatibility Pass 2011/65/EU

Specifications

Format Manual
Product Type Lockout Compliance Manual
For Use With (Application) Step-by-step instructions for developing a hazardous energy control program compliance
Subject Safety and Training Materials