missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Keyed Different Safety Padlocks with 1.5 in. Shackle, Red

Catalog No. 19000308
Change view
Click to view available options
Quantity:
6/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19000308 6/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19000308 Supplier Brady™ Supplier No. 51339

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Keyed-Different Safety Padlocks with 1.5 in. Shackle, Red is a safer alternative to metal locks for lockout applications.

  • Padlocks feature lightweight, nonconductive fiberglass-reinforced nylon bodies, hardened steel shackles and tamper-resistant 6-pin cylinders
  • These lightweight padlocks comply with all OSHA requirements and offer superior temperature, chemical and corrosion-resistance
  • Non-conductive safety padlocks are a preferred alternative to metal locks for lockout applications
  • Each padlock is keyed differently with keys precision-machined to exacting standards, so that the key for one lock will not open another in the set

Specifications

Product Type Keyed Different Safety Padlock
Color Red
Material Nylon
Certifications/Compliance OSHA 1910.147
Diameter (English) Shackle 0.25 in.
Diameter (Metric) Shackle 0.63 cm
For Use With (Equipment) Industrial Manufacturing; Food and Beverage
Includes 3 labels/ lock, 1 English, 1 French, 1 Spanish label
Material (Shackle) Steel
No. of Pins 6
Quantity 6/Pk.
Show More Show Less