Learn More
Brady™ Keyed-Different Nylon Safety Padlocks with 1.5 in. Shackle Locks
Padlock offers unique keyway which breaks any current traveling form padlock shackle to cylinder
Supplier: Brady™ 123356
Description
- Keyed-different padlock with non-conductive, spark resistant lock body
- Compact and lightweight
- Available in a variety of colors
Specifications
| Black | |
| Keyed Different | |
| Nylon | |
| 6/Pk. |