Learn More
Brady™ Keyed-Different Nylon Safety Padlocks with 1.5 in. Shackle Locks
Padlock offers unique keyway which breaks any current traveling form padlock shackle to cylinder
Supplier:  Brady™ 123352
 View more versions of this product
Catalog No. 19-103-471
Only null left
Description
Description
  • Keyed-different padlock with non-conductive, spark resistant lock body
  • Compact and lightweight
  • Available in a variety of colors
Specifications
Specifications
Blue
Keyed Different
Nylon
6/Pk.
Product Suggestions
Product Suggestions