Learn More
Brady™ Keyed-Alike Safety Locks
Keyed-Alike Safety Lock padlocks from Brady™ have nonconductive fiberglass-reinforced nylon bodies and unique key cylinders that prevent electricity from traveling from the shackle to the key.
Supplier:  Brady™ 118968
 View more versions of this product
Catalog No. 19140372
Only null left
Description
Description
  • Extremely compact and lightweight - compact safety padlocks from Brady™ are the smallest safety locks on the market
  • These lightweight padlocks comply with all OSHA requirements and offer superior temperature, chemical and corrosion-resistance
  • Non-conductive safety padlocks are a preferred alternative to metal locks for lockout applications
  • Every lock in this set can be opened with the same key, which is beneficial when multiple locks are assigned to a single employee
Specifications
Chemical Identifiers
RoHs, ANSI Z244, OSHA 29 CFR 1910.147
1 in.
2.54 cm
0.6 cm
1.52 cm
Key and Labels in English, Spanish and French, Special 5-pin Cylinder
Compact Safety Padlock
1.4 in.
3 cm
1.2 in.
0.6 in.
0.25 in.
Keyed Alike
3.6 cm
Plastic
For Certain Lockout Applications
Electrical; Confined Spaces
Specifications
RoHs, ANSI Z244, OSHA 29 CFR 1910.147
1 in.
2.54 cm
Purple
Keyed Alike
3.6 cm
Plastic
Compact Safety Padlock
1.4 in.
3 cm
1.2 in.
0.6 in.
0.25 in.
0.6 cm
1.52 cm
Key and Labels in English, Spanish and French, Special 5-pin Cylinder
6/Pk.
For Certain Lockout Applications
Electrical; Confined Spaces
Product Suggestions
Product Suggestions