Learn More
Brady™ Keyed Alike Nylon Safety Padlocks with 1.5 in. Shackle Locks
Catalog No. 19103469
Change view
Click to view available options
Color:
Brown
Green
Orange
Purple
White
Quantity:
3/Pk.
6/Pk.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Quantity
19103469 White 6/Pk.
19103454 Green 3/Pk.
19103465 Orange 6/Pk.
19103467 Brown 6/Pk.
19103468 Purple 6/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19103469 Supplier Brady™ Supplier No. 123350
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Padlock offers unique keyway which breaks any current traveling form padlock shackle to cylinder
  •  Keyed-Alike padlock with non-conductive, spark resistant lock body
  • Compact and lightweight
  • Available in a variety of colors
Specifications
Color White
Key Type Keyed Alike
Material Nylon
Quantity 6/Pk.
Product Type Padlock
Width (English) Shackle 1.5 in.