Learn More
Brady™ Keyed Alike Nylon Safety Padlocks with 1.5 in. Shackle Locks
Padlock offers unique keyway which breaks any current traveling form padlock shackle to cylinder
Supplier: Brady™ 123347
Description
- Keyed-Alike padlock with non-conductive, spark resistant lock body
- Compact and lightweight
- Available in a variety of colors
Specifications
| Black | |
| Nylon | |
| Padlock |
| Keyed Alike | |
| 6/Pk. | |
| 1.5 in. |