Learn More
Brady™ Keyed Alike Nylon Safety Padlocks with 1.5 in. Shackle Locks
Padlock offers unique keyway which breaks any current traveling form padlock shackle to cylinder
Supplier:  Brady™ 123343
 View more versions of this product
Catalog No. 19103462
Only null left
Description
Description
  •  Keyed-Alike padlock with non-conductive, spark resistant lock body
  • Compact and lightweight
  • Available in a variety of colors
Specifications
Chemical Identifiers
Keyed Alike
Nylon
Padlock
1.5 in.
Specifications
Blue
Nylon
Padlock
Keyed Alike
6/Pk.
1.5 in.
Product Suggestions
Product Suggestions