Learn More
Brady™ Keyed Alike Nylon Safety Padlocks with 1.5 in. Shackle Locks
Catalog No. 19103453
Change view
Click to view available options
Color:
Black
Blue
Brown
Green
Orange
Purple
White
Quantity:
3/Pk.
6/Pk.
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Quantity
19103453 Blue 3/Pk.
19103454 Green 3/Pk.
19103457 Black 3/Pk.
19103459 Purple 3/Pk.
19103462 Blue 6/Pk.
19103463 Green 6/Pk.
19103465 Orange 6/Pk.
19103467 Brown 6/Pk.
19103468 Purple 6/Pk.
19103469 White 6/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 19103453 Supplier Brady™ Supplier No. 123334
Padlock offers unique keyway which breaks any current traveling form padlock shackle to cylinder
  •  Keyed-Alike padlock with non-conductive, spark resistant lock body
  • Compact and lightweight
  • Available in a variety of colors
Specifications
Color Blue
Key Type Keyed Alike
Material Nylon
Quantity 3/Pk.
Product Type Padlock
Width (English) Shackle 1.5 in.