missing translation for 'onlineSavingsMsg'
Learn More

Brady™ HMIG Labels

Catalog No. 19823719
Change view
Click to view available options
Color:
Blue, Black, Yellow, White, Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19823719 Blue, Black, Yellow, White, Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19823719 Supplier Brady™ Supplier No. 60325

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

HMIG Labels are made with Coated Paper (B-235), which features surface printed graphics on semi-gloss paper label stock, making it ideal for general label applications.

  • Label contains: CHEMICAL NAME, COMMON NAME, MANUFACTURER, DATE
  • B-235 Coated Paper with pressure sensitive acrylic adhesive adheres to most surfaces
  • Labels remove cleanly, leaving no residue on surface
  • Recommended for indoor use

Specifications

Autoclavable Not Autoclavable
Certifications/Compliance Globally Harmonized System of Classification and Labeling of Chemicals (GHS), NFPA 704, OSHA 1910.1200
Description Label
Adhesive Type Permanent Acrylic
Color Blue, Black, Yellow, White, Red
Finish Semi-gloss
Format Label
Material Paper
Printable Sides Single-side Printable
Product Type Hazardous Materials Label
Sterility Non-sterile
For Use With (Application) Facility Identification, Hazard Communication Identification, Safety Identification
Includes Roll of 1000 Labels
Languages English
Quantity 1000/Roll
Shape Rectangle
Thickness (English) 0.0038 in.
Dimensions (L x W) 1.5 x 3.875 in.
Length (English) 1.5 in.
Width (English) 3.875 in.
Width (Metric) 98.43 mm
Height (English) 1.5 in.
Height (Metric) 38.10 mm
Show More Show Less