Learn More
Brady™ High Visibility Signs - Flexible
Flexible High Visibility Signs are made of flexible polyethylene (B-450), which is ideal for high visibility fire equipment signs and a variety of custom markers and signs.
Supplier:  Brady™ 49043
 View more versions of this product
Catalog No. 19054834
Only null left
Description
Description
  • V-style 45° high visibility signs project from the wall to be easily viewed from all angles
  • 3 to 5 year average outdoor durability
  • Linear polyethylene sheet contains UV stabilizers
  • Surface printing holds up to most common chemicals, oils and water
Specifications
Chemical Identifiers
ASTM D 1593, ASTM D 638, ASTM D 635
Polyethylene
3600 psi
179.6°F
0.06 in.
45.72 cm
-40°F to 179.6°F
High Visibility Sign
Industrial
1 Ea.
12 in.
-40°F
18 in.
-40°C to 82°C
12 x 18 in.
Specifications
ASTM D 1593, ASTM D 638, ASTM D 635
Polyethylene
3600 psi
12 in.
-40°F
18 in.
-40°C to 82°C
12 x 18 in.
High Visibility Sign
Industrial
1 Ea.
Non-adhesive
179.6°F
0.06 in.
45.72 cm
-40°F to 179.6°F
Safety Shower
Green on White
Product Suggestions
Product Suggestions