missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Hazwik™ Spill Buckets
Description
- Surfactant-treated, brightly colored absorbent picks up a wide range of aggressive chemicals while remaining chemically inert
- 6.5-Gallon Bucket is ideal for small spill response, is easily accessible, and reusable
- Improve worker productivity and efficiency by placing spill kits around your facility close to areas prone to spills
- Spill kits are ideal for fast response to critical accidents
Specifications
Specifications
| Color | White |
| Capacity (English) | 6.5 gal. |
| Capacity (Metric) | 24.6 L |
| Material | Polyethylene |
| Type | Bucket Spill Kit |
| For Use With (Application) | Small spill response of hazardous materials, for oil based material |
| Includes | (10) 15 x 19 in. pads, (3) 3 in. x 4 ft. SOCs, 17 x 19 in. pillows, 1 pair nitrile gloves, 2 disposal bags, instruction sheet |
| Packaging Quantity | 1 Ea. |
| Length (English) | 18 in. |
| Length (Metric) | 45.72 cm |
| Show More |