missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Hazardous Material Shipping Labels

Catalog No. 19807138
Change view
Click to view available options
Color:
Orange, White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19807138 Orange, White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19807138 Supplier Brady™ Supplier No. 63242

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ETIOLOGIC AGENTS—BIOMEDICAL MATERIAL: IN CASE OF DAMAGE OR… hazardous materials shipping label meets DOT requirements (reglation HM-181).

  • Made of orange plastic
  • Size: 2 in H x 4 in W
  • Self adhesive

Specifications

Certifications/Compliance DOT HM-181
Adhesive Type Permanent, High-tack
Color Orange, White
Finish Matte
Format Label
Material Composite Plastic
Product Type Label
Sterility Non-sterile
For Use With (Application) Hazardous material label
Quantity 500/Pk.
Temperature Range (English) -50 to 150°F
Temperature Range (Metric) -45 to 65.5°C
Dimensions (L x W) 2 x 4 in.
Length (English) 2 in.
Width (English) 4 in.
Width (Metric) 101.60 mm
Height (English) 2 in.
Height (Metric) 5.08 cm
Show More Show Less