Learn More
Brady™ GlobalMark™ Polyester Tapes
Durable outdoor-grade polyester with permanent adhesive.
Supplier:  Brady™ 76646
 View more versions of this product
Catalog No. 19201175
Only null left
Description
Description
  • Low-halide content; ideal for marking stainless-steel pipes and surfaces
  • Ultrasmooth finish is excellent for mounting to rigid panels and very smooth surfaces
  • Two-year average outdoor durability in temperatures from 110°C to -40°C (230°F to -40°F); can be applied in temperatures as low as -18°C (0°F)
Order Info
Thickness: 3mil
Specifications
Chemical Identifiers
Tape
Ideal for Stainless Steel and surfaces
-40°C to +110°C
Non-sterile
-40°F to 230°F
Specifications
Acrylic, Permanent, Pressure-sensitive
Consignment
Tape
Single-side Printable
Label Tape
Ideal for Stainless Steel and surfaces
80 Gardner Units
1 Ea.
-40°C to +110°C
100 ft. x 0.5 in.
30.48 m
12.7 mm
Brown
Gloss
Polyester
Thermal Transfer
Non-sterile
GlobalMark™ 2
2 Years
-40°F to 230°F
3 mil
100 ft.
0.5 in.
Product Suggestions
Product Suggestions