missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Globally Harmonized System (GHS) Flammable Picto Labels

Catalog No. 19140549
Change view
Click to view available options
Quantity:
16/Pk.
39/Pk.
4/Pk.
Material:
Coated Paper
Vinyl
Height (English):
0.62 in.
1 in.
2 in.
Width (English):
0.6 in.
1 in.
2 in.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Material Height (English) Width (English)
19140549 4/Pk. Vinyl 2 in. 2 in.
19140513 39/Pk. Vinyl 0.62 in. 0.6 in.
19140522 39/Pk. Coated Paper 0.62 in. 0.6 in.
19140531 16/Pk. Vinyl 1 in. 1 in.
19140540 16/Pk. Coated Paper 1 in. 1 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19140549 Supplier Brady™ Supplier No. 118823
Only null left

For flammable gases, liquids and solids; pyrophorics; self-reatives; self-heating

Pre-printed die cut labels identify hazardous products for chemical risks, health risks and environmental risks in accordance to the United Nations' universal approach to classifying and communicating chemical hazards.

  • Legend: Flammable
  • Made of Vinyl or Coated paper
  • Available in three sizes

Specifications

Certifications/Compliance GHS, OSHA 29 CFR 1910.1200
Adhesive Type Permanent Acrylic
Color Black, Red, White
Finish Gloss
Format Label
Material Vinyl
Product Type Pictogram Label
Sterility Non-sterile
Display Text Flammable
For Use With (Application) Flammable gases, liquids and solids
Quantity 4/Pk.
Temperature Range (English) -20°F to 75°F
Temperature Range (Metric) -4°C to 167°C
Dimensions (L x W) 2 x 2 in.
Width (English) 2 in.
Height (English) 2 in.
Height (Metric) 5 cm
Show More Show Less