missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Globally Harmonized System (GHS) Explosive Picto Labels
Description
Pre-printed die cut labels identify hazardous products for chemical risks, health risks and environmental risks in accordance to the United Nations' universal approach to classifying and communicating chemical hazards.
- Legend: Explosive
- Made of Vinyl or Coated paper
- Available in three sizes
Specifications
Specifications
| Adhesive Type | Acrylic, Permanent |
| Color | Black, Red, White |
| Material | Vinyl |
| Product Type | Pictogram Label |
| For Use With (Application) | Explosives, Self-reactives, Organic Peroxides |
| Life Span | 3 Years |
| Quantity | 4/Pk. |
| Temperature Range (English) | -20°F to 75°F |
| Temperature Range (Metric) | -4°C to 167°C |
| Thickness (English) | 0.105 in. |
| Show More |