Learn More
Brady™ Globally Harmonized System (GHS) Explosive Picto Labels
For explosive, self-reactive, organic peroxides
Supplier:  Brady™ 118870
 View more versions of this product
Catalog No. 19140539
Only null left
Description
Description
Pre-printed die cut labels identify hazardous products for chemical risks, health risks and environmental risks in accordance to the United Nations' universal approach to classifying and communicating chemical hazards.
  • Legend: Explosive
  • Made of Vinyl or Coated paper
  • Available in three sizes
Specifications
Chemical Identifiers
Black, Red, White
Explosives, Self-reactives, Organic Peroxides
Specifications
Acrylic
Semi-gloss
Coated Paper
Non-sterile
16/Pk.
-40°C to +65°C
1 x 1 in.
1 in.
Black, Red, White
Label
Pictogram Label
Explosives, Self-reactives, Organic Peroxides
-40°F to 150°F
0.0038 in.
1 in.
2.54 cm
Product Suggestions
Product Suggestions