missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Globally Harmonized System (GHS) Explosive Picto Labels
For explosive, self-reactive, organic peroxides
Supplier: Brady™ 118816
Description
Pre-printed die cut labels identify hazardous products for chemical risks, health risks and environmental risks in accordance to the United Nations' universal approach to classifying and communicating chemical hazards.
- Legend: Explosive
- Made of Vinyl or Coated paper
- Available in three sizes
Chemical Identifiers
| Black, Red, White | |
| Explosives, Self-reactives, Organic Peroxides |
Specifications
| Acrylic, Permanent | |
| Gloss | |
| Vinyl | |
| Non-sterile | |
| 3 Years | |
| -20°F to 75°F | |
| 0.004 in. | |
| 1 in. | |
| 2.54 cm |
| Black, Red, White | |
| Label | |
| Pictogram Label | |
| Explosives, Self-reactives, Organic Peroxides | |
| 16/Pk. | |
| -4°C to 167°C | |
| 1 x 1 in. | |
| 1 in. |