missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Globally Harmonized System (GHS) Explosive Picto Labels
For explosive, self-reactive, organic peroxides
Supplier: Brady™ 118869
Description
Pre-printed die cut labels identify hazardous products for chemical risks, health risks and environmental risks in accordance to the United Nations' universal approach to classifying and communicating chemical hazards.
- Legend: Explosive
- Made of Vinyl or Coated paper
- Available in three sizes
Chemical Identifiers
| Black, Red, White | |
| Explosives, Self-reactives, Organic Peroxides |
Specifications
| Acrylic | |
| Semi-gloss | |
| Coated Paper | |
| Non-sterile | |
| 39/Pk. | |
| -40°C to +65°C | |
| 0.625 x 0.625 in. | |
| 0.62 in. |
| Black, Red, White | |
| Label | |
| Pictogram Label | |
| Explosives, Self-reactives, Organic Peroxides | |
| -40°F to 150°F | |
| 0.0038 in. | |
| 0.6 in. | |
| 15.7 mm |