Learn More
Brady™ Globally Harmonized System (GHS) Acute Toxic Picto Labels
For acute toxicity, skin and eye irritation, skin sensitizers
Supplier:  Brady™ 118859
 View more versions of this product
Catalog No. 19-140-706
Only null left
Explore available promotions
Description
Description
Pre-printed die cut labels identify hazardous products for chemical risks, health risks and environmental risks in accordance to the United Nations' universal approach to classifying and communicating chemical hazards
  • Legend: Acute Toxic
  • Made of B-7569 Viny or B-235 Paper
  • Available in three sizes
Specifications
Specifications
Acrylic, Pressure-sensitive
Gloss
Vinyl (B-7569)
Acute Toxic
4/Card
5.08 x 5.08 cm
2 in.
Black, Red, White
Label
Non-sterile
For acute toxicity, skin and eye irritation, skin sensitizers
2 x 2 in.
2 x 2 in.
Product Suggestions
Product Suggestions