missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Globally Harmonized System (GHS) Acute Toxic Picto Labels
For acute toxicity, skin and eye irritation, skin sensitizers
Supplier: Brady™ 118911
Description
Pre-printed die cut labels identify hazardous products for chemical risks, health risks and environmental risks in accordance to the United Nations' universal approach to classifying and communicating chemical hazards
- Legend: Acute Toxic
- Made of B-7569 Viny or B-235 Paper
- Available in three sizes
Specifications
| GHS, OSHA 29 CFR 1910.1200 | |
| Black, Red, White | |
| Label | |
| Pictogram Label | |
| Acute Toxic | |
| 39/Pk. | |
| 1.58 x 1.58 cm (W x H) | |
| -40°C to +65°C | |
| 0.625 x 0.625 in. | |
| 0.62 in. |
| Acrylic, Pressure-sensitive | |
| Semi-gloss | |
| Paper | |
| Non-sterile | |
| For acute toxicity, skin and eye irritation, skin sensitizers | |
| 0.625 x 0.625 in. | |
| -40°F to 150°F | |
| 0.0038 in. | |
| 0.6 in. | |
| 15.20 mm |