missing translation for 'onlineSavingsMsg'
Learn More

Brady™ General Traffic Signs

Brady traffic signs provide direction or information to sustain a safe work environment.

Supplier:  Brady™ 75207

 View more versions of this product

Catalog No. 19807239


Only null left

Description

Description

  • Durable Reflective Aluminum (B-959) is constructed of reflective sheeting applied to an aluminum sign panel and is ideal for traffic control applications.
  • Extremely durable (B-120) Premium Fiberglass will not chip, fade, rust, shatter or peel for up to 25 years.
Specifications

Chemical Identifiers

1 Ea.
Non-adhesive
-40°F
Traffic Sign

Specifications

MUTCD
1 Ea.
Non-adhesive
Industrial, utility, commercial and institutional environment
193°F
0.1 in.
76.2 cm
-40°F to 193°F
Stop
Red on White
Fiberglass
9000 psi
Premium Fiberglass
30 in.
-40°F
30 in.
-40°C to 90°C
30 x 30 in.
Traffic Sign
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions