Learn More
Brady™ General and Industrial BBP33 Labels: Three Per Row
Wire and Cable Identification BBP33 Labels (Three per Row) are polyester or polypropylene wrap-around labels that are highly pliable to conform to tightly curved surfaces such as wire and cable.
Supplier:  Brady™ B335423
 View more versions of this product
Catalog No. 19140562
Only null left
Description
Description
  • Self-Laminating Polyester (B-461) has a permanent acrylic adhesive and is thermal transfer printable, good print smudge resistance, solvent resistance and high/low temperature performance. Glossy white finish with permanent acrylic (B-423) meets requirements of a halogen-free material per DIN VDE 0472 part 815 and has a service temperature of -94 to 230° F (-70 to 110° C).
  • B-425 thermal transfer printable Polypropylene is ideal for labeling applications requiring excellent solvent resistance and print performance.
Specifications
Chemical Identifiers
Acrylic, Permanent
Thermal Transfer
Non-sterile
BBP™ 33 Printer
Specifications
CSA, RoHS, UL
Acrylic, Permanent
Consignment
Die-cut
Single-side Printable
Industrial Label
Electronic PCB and component identification, Bar code label, rating plates and solar panel identification
27.31 mm
-70°C
5000/Pk.
LabelMark, MarkWare
-70°C to +120°C
0.076 mm
0.5 x 1 in.
1 in.
85.09 mm
12.70 mm
Polyester Labels for the BBP33 Printer
White
Gloss
Polyester
Thermal Transfer
Non-sterile
BBP™ 33 Printer
3
5000 Labels/Box
Rectangle
-94°F to 248°F
0.003 in.
15.24 mm
0.5 in.
25.4 mm
0.5 in.
B33-5-423
Product Suggestions
Product Suggestions