missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Floor Safety Signs
Durable, highly visible Floor Safety Signs are designed to warn of hazardous areas, putting valuable safety information at the point of need.
$62.00 - $63.19
Specifications
| Material | Vinyl |
|---|---|
| Quantity | 1 Ea. |
| Adhesive Type | Permanent Acrylic |
| Diameter (English) | 17 in. |
| For Use With (Application) | Designed to warn of hazardous areas, putting valuable safety information at the point of need |
| Catalog Number | Mfr. No. | Color | Displayed Text | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Displayed Text | Price | Quantity & Availability | ||||
|
19054837
|
Brady™
49064 |
Red on White | Fire Extinguisher |
Each for $63.19
|
|
||||
|
19054840
|
Brady™
49067 |
Red on White | Emergency Exit |
Each for $62.20
|
|
||||
|
19054888
|
Brady™
92404 |
Black on Yellow | Watch Out For Forklift Trucks |
Each for $62.00
|
|
||||
Description
- Floor safety signs improve facility safety by alerting employees to hazards and/or important safety information in areas with foot traffic
- B-819 Anti-Skid Vinyl features a velvet embossed rigid overlaminate
- Excellent chemical, oil and water resistance
- Minimum application temperature of 0°F (-18°C)
Specifications
| Vinyl | |
| Permanent Acrylic | |
| Designed to warn of hazardous areas, putting valuable safety information at the point of need | |
| -40°F | |
| -40°C to 70°C | |
| Floor Safety Sign |
| 1 Ea. | |
| 17 in. | |
| 158°F | |
| 0.01 in. | |
| -40°F to 158°F |