missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Empty Cylinders Sign

Catalog No. 19016631
Change view
Click to view available options
Material:
Fiberglass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19016631 Fiberglass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19016631 Supplier Brady™ Supplier No. 70317

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Identify equipment, rooms and materials that contain or are contaminated with hazardous agents.

  • Bright colors and bold text provide high visibility to critical messages
  • Sign reads EMPTY CYLINDERS
  • Designed to withstand harsh industrial environments
  • White on blue
  • Size: 25.4H × 35.56cmW (10H × 14 in.W)
  • B-120 premium fiberglass — fiberglass reinforced polyester

Specifications

Material Fiberglass
Quantity 1 Ea.
Tensile Strength 9000 psi
Description Premium Fiberglass
Mounting Type Surface Mount
For Use With (Application) Industrial, utility, commercial and institutional environment
Height (English) 10 in.
Max. Temperature (English) 193°F
Min. Temperature (English) -40°F
Thickness 0.1 in.
Width (English) 14 in.
Temperature Range (Metric) -40°C to 90°C
Shape Rectangular
Temperature Range (English) -40°F to 193°F
Languages English
Displayed Text Empty Cylinders
Product Type Chemical and Hazardous Materials Sign
Color White on Blue
Show More Show Less