missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Electrical Hazard Signs: WARNING - HIGH VOLTAGE
Identify electrical hazard and high voltage areas
$47.33
Specifications
| Certifications/Compliance | ANSI Z535, OSHA 1910.145, PSTC-33, |
|---|---|
| Quantity | 1 Ea. |
| Mounting Type | Surface Mount |
| Height (English) | 10 in. |
| Height (Metric) | 25.40 cm |
Description
- Available in two materials - plastic and fiberglass
- Black on orange
Specifications
| ANSI Z535, OSHA 1910.145, PSTC-33, | |
| Surface Mount | |
| 25.40 cm | |
| 14 in. | |
| Warning High Voltage |
| 1 Ea. | |
| 10 in. | |
| -40°F | |
| 35.56 cm | |
| Electrical Hazard Sign |