missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Economy-To-Go Pipe Markers
Description
- Pipe Markers-To-Go are made of economical B-736 surface printed plastic for use on smooth, clean, dry pipe surfaces.
- Markers withstand temperatures from -40 degree F to 212 degree F.
- Supplied in rolls of 25 identical markers and matching arrows.
Specifications
Specifications
| Certifications/Compliance | ANSI A13.1 1981 |
| Description | Pipe Marker |
| Adhesive Type | Permanent, Pressure-sensitive |
| Color | Black, Yellow |
| Material | Plastic |
| Sterility | Non-sterile |
| Display Text | Refrigerated Suction |
| For Use With (Application) | Ideal for use on clean, dry pipes, each marker has a diecut design which enables each label and arrow to be applied in the selected flow directions |
| For Use With (Equipment) | Pipe & Valve Marking |
| Gloss | 80 to 95 Gardner Units |
| Show More |