Learn More
Brady™ Dash Space
Dash Space B-514 polyester material is ideal for use in marking aisles, passageways, storage locations, and point out safety equipment and exits.
Supplier:  Brady™ 142179
 View more versions of this product
Catalog No. 19150900
Only null left
Description
Description
  • High-gloss surface shines like new paint and withstands forklift traffic without tearing or lifting
  • Tape removes cleanly without any unsightly adhesive residue or damage to the floor
  • Ideal for marking aisles, passageways and storage locations and identifying equipment and egress paths
  • Exceeds safety standards for clean, dry non-slip surfaces per ANSI A 1264.2-2006 and OSHA 1910.32
Specifications
Chemical Identifiers
RoHS
46/Pk.
ToughStripe Prespaced shapes
0.008 in.
50.8 mm
Label
Polyester
Rubber-based
2 in.
8 in.
-18 °C-54 °C
Specifications
RoHS
46/Pk.
ToughStripe Prespaced shapes
0.008 in.
50.8 mm
Label
Polyester
Rubber-based
2 in.
8 in.
-18 °C-54 °C
Red
Product Suggestions
Product Suggestions