missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Danger: Wear Face Shield And Rubber Gloves... Signs

Catalog No. 19036530
Change view
Click to view available options
Material:
Fiberglass
Plastic
Height (English):
7 in.
10 in.
Width (English):
10 in.
14 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Height (English) Width (English)
19036530 Plastic 7 in. 10 in.
1704619 Fiberglass 10 in. 14 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19036530 Supplier Brady™ Supplier No. 25222

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Identify equipment, rooms and materials that contain or are contaminated with hazardous agents.

  • Bright colors and bold text provide high visibility to critical messages
  • Sign reads “DANGER - WEAR FACE SHIELD AND RUBBER GLOVES WHEN HANDLING CHEMICALS”
  • Black/red on white
  • Available in different sizes and materials

Specifications

Certifications/Compliance ASTM D 1593, ANSI Z535, OSHA 1910.145
Material Plastic
Quantity 1 Ea.
Mounting Type Surface Mount
For Use With (Application) Light duty industrial, utility, commercial and institutional environments for both indoor and outdoor use
Height (English) 7 in.
Height (Metric) 17.7 cm
Max. Temperature (English) 175°F
Min. Temperature (English) -40°F
Thickness 0.06 in.
Width (English) 10 in.
Width (Metric) 17.7 cm
Temperature Range (Metric) -40°C to 80°C
Shape Rectangular
Temperature Range (English) -40°F to 175°F
Languages English
Displayed Text DANGER Wear Face Shield And Rubber Gloves When Handling Chemicals
Product Type Protective Wear Sign
Color Red on White
Show More Show Less