missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Danger: Nitrogen Signs

Catalog No. 19102054
Change view
Click to view available options
Material:
Plastic
Polyester
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19102054 Plastic
19035719 Polyester
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19102054 Supplier Brady™ Supplier No. 22332

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Identifying equipment, rooms and materials that contain or are contaminated with hazardous agents

  • Bright colors and bold text provide high visibility to critical messages
  • Sign reads “DANGER NITROGEN”
  • Black/red on white
  • Designed to withstand harsh industrial environments
  • Available in different sizes and materials

B-401

  • Plastic — high-impact polystyrene
  • B-302
    • Self-sticking polyester — surface printed polyester, polyester overlaminate and sensitive acrylic

    Specifications

    Certifications/Compliance ANSI Z535, OSHA 1910.145
    Material Plastic
    Quantity 1 Ea.
    Mounting Type Surface Mount
    For Use With (Application) Light duty industrial, utility, commercial, institutional environments
    Height (English) 10 in.
    Height (Metric) 25.4 cm
    Max. Temperature (English) 175°F
    Min. Temperature (English) -40°F
    Thickness 0.06 in.
    Width (English) 14 in.
    Width (Metric) 35.56 cm
    Temperature Range (Metric) -40°C to 80°C
    Shape Rectangular
    Temperature Range (English) -40°F to 175°F
    Languages English
    Displayed Text DANGER Nitrogen
    Product Type Chemical and Hazardous Materials Sign
    Color Red on White
    Show More Show Less