missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Danger: Hazardous Waste Storage Area… Signs

Identify equipment, rooms and materials that contain or are contaminated with hazardous agents

Supplier:  Brady™ 84080

 View more versions of this product

Catalog No. 19035598


Only null left

Description

Description

  • Sign reads “DANGER HAZARDOUS WASTE STORAGE AREA UNAUTHORIZED PERSONS KEEP OUT”
  • Black/red on white
  • Designed to withstand harsh industrial environments
  • Available in different sizes and materials

B-401

  • Plastic — high-impact polystyrene
  • B-120
    • Premium fiberglass — fiberglass reinforced polyester
  • B-302
    • Self-sticking polyester — surface printed polyester, polyester overlaminate and sensitive acrylic
  • B-555
    • Aluminum — 0.040 in. coil coated aluminum construction

    Specifications

    Chemical Identifiers

    1 Ea.
    -40°F
    Admittance Sign
    Surface Mount
    DANGER Hazardous Waste Storage Area Unauthorized Persons Keep Out
    Red on White

    Specifications

    ANSI Z535, OSHA 1910.145
    1 Ea.
    Identification of fire alarms, extinguishers and emergency exits
    17.7 cm
    -40°F
    10 in.
    -40°C to 110°C
    DANGER Hazardous Waste Storage Area Unauthorized Persons Keep Out
    Red on White
    Polyester
    Surface Mount
    7 in.
    230°F
    0.006 to 0.01 in.
    25.4 cm
    -40°F to 230°F
    Admittance Sign
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Promotions

    Promotions