missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Danger: Hazardous Waste Storage Area… Signs

Catalog No. 19035597
Change view
Click to view available options
Material:
Fiberglass
Plastic
Polyester
Width (English):
10 in.
14 in.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Width (English)
19035597 Plastic 14 in.
19035596 Plastic 10 in.
19016660 Fiberglass 14 in.
19035598 Polyester 10 in.
19035599 Polyester 14 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19035597 Supplier Brady™ Supplier No. BRD22102

Identify equipment, rooms and materials that contain or are contaminated with hazardous agents

  • Sign reads “DANGER HAZARDOUS WASTE STORAGE AREA UNAUTHORIZED PERSONS KEEP OUT”
  • Black/red on white
  • Designed to withstand harsh industrial environments
  • Available in different sizes and materials

B-401

  • Plastic — high-impact polystyrene
  • B-120
    • Premium fiberglass — fiberglass reinforced polyester
  • B-302
    • Self-sticking polyester — surface printed polyester, polyester overlaminate and sensitive acrylic
  • B-555
    • Aluminum — 0.040 in. coil coated aluminum construction

    Specifications

    Certifications/Compliance ANSI Z535, OSHA 1910.145, ISO 9000, ASTM D 1593
    Material Plastic
    Quantity 1 Ea.
    Mounting Type Surface Mount
    For Use With (Application) Light duty industrial, utility, commercial, institutional environments
    Height (English) 10 in.
    Height (Metric) 25.4 cm
    Max. Temperature (English) 175°F
    Min. Temperature (English) -40°F
    Thickness 0.06 in.
    Width (English) 14 in.
    Width (Metric) 35.5 cm
    Temperature Range (Metric) -40°C to 80°C
    Temperature Range (English) -40°F to 175°F
    Displayed Text DANGER Hazardous Waste Storage Area Unauthorized Persons Keep Out
    Product Type Admittance Sign
    Color Red on White
    Show More Show Less