Learn More
Brady™ Danger: Hazardous Waste Signs
For identifying equipment, rooms and materials that contain or are contaminated with hazardous agents
Supplier:  Brady™ 22753
 View more versions of this product
Catalog No. 19035713
Only null left
Description
Description
  • Bright colors and bold text provide high visibility to critical messages
  • Sign reads DANGER HAZARDOUS WASTE
  • Black/red on white
  • Designed to withstand harsh industrial environments
  • Available in different sizes and materials
B-401
  • Plastic — high-impact polystyrene
  • B-302
    • Self-sticking polyester — surface printed polyester, polyester overlaminate and sensitive acrylic
    Specifications
    Chemical Identifiers
    1 Ea.
    -40°F
    Chemical and Hazardous Materials Sign
    Surface Mount
    DANGER Hazardous Waste
    Specifications
    ANSI Z535, OSHA 1910.145, ASTM D 1593
    1 Ea.
    Light duty industrial, utility, commercial, institutional environments
    17.78 cm
    -40°F
    10 in.
    -40°C to 80°C
    DANGER Hazardous Waste
    Black
    Plastic
    Surface Mount
    7 in.
    175°F
    0.06 in.
    25.4 cm
    -40°F to 175°F
    Chemical and Hazardous Materials Sign
    Product Suggestions
    Product Suggestions