missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Danger Hazardous Area Signs
Danger: Hazardous Area signs (B-302) are high-performance, pressure-sensitive safety signs constructed of surface-printed polyester with a polyester overlaminate.
$13.41 - $14.53
Specifications
| Grade | Industrial |
|---|---|
| Material | Polyester |
| Quantity | 1 Ea. |
| Adhesive Type | Permanent Acrylic |
| For Use With (Application) | Identification of fire alarms, extinguishers and emergency exits |
| Catalog Number | Mfr. No. | Displayed Text | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Displayed Text | Price | Quantity | |||||
|
19-823-253
|
Brady™
85493 |
DANGER Hazardous Area |
Each for $14.53
|
|
|||||
|
19823254
|
Brady™
85497 |
DANGER Hazardous Materials |
Each for $13.41
|
|
|||||
Description
- Weathers up to 8 years outdoors, even longer indoors, and withstands wide temperature variations (up to 230°F)
- Flexible material conforms to most surfaces while protected graphics hold up to harsh environments, abrasion, and repeated spills and cleanings
- Permanent, pressure sensitive adhesive forms a long-lasting bond to even the most difficult surfaces and shapes
- Cold temperature adhesive allows the signage to be applied at 0°F (-18°C)
Specifications
| Industrial | |
| 1 Ea. | |
| Identification of fire alarms, extinguishers and emergency exits | |
| 230°F | |
| 0.006 to 0.01 in. | |
| 25.4 cm | |
| -40°F to 230°F | |
| Chemical and Hazardous Materials Sign |
| Polyester | |
| Permanent Acrylic | |
| 7 in. | |
| -40°F | |
| 10 in. | |
| -40°C to 110°C | |
| 7 x 10 in. | |
| Red on White |