missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Danger: Corrosive Signs

Catalog No. 19035704
Change view
Click to view available options
Material:
Fiberglass
Plastic
Polyester
Height (English):
7 in.
10 in.
Width (English):
10 in.
14 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Height (English) Width (English)
19035704 Plastic 7 in. 10 in.
1704612 Fiberglass 10 in. 14 in.
19035705 Plastic 10 in. 14 in.
19035706 Polyester 10 in. 14 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19035704 Supplier Brady™ Supplier No. BRD22323

Identify equipment, rooms and materials that contain or are contaminated with hazardous agents.

  • Bright colors and bold text provide high visibility to critical messages
  • Sign reads DANGER CORROSIVE
  • Black/red on white
  • Designed to withstand harsh industrial environments
  • Available in different sizes and materials

B-401

  • Plastic — high-impact polystyrene
  • B-120
    • Premium fiberglass — fiberglass reinforced polyester
  • B-302
    • Self-sticking polyester — surface printed polyester, polyester overlaminate and sensitive acrylic

    Specifications

    Certifications/Compliance ANSI Z535, OSHA 1910.145, ASTM D 1593
    Material Plastic
    Quantity 1 Ea.
    Mounting Type Surface Mount
    For Use With (Application) Light duty industrial, utility, commercial, institutional environments
    Height (English) 7 in.
    Height (Metric) 17.7 cm
    Max. Temperature (English) 175°F
    Min. Temperature (English) -40°F
    Thickness 0.06 in.
    Width (English) 10 in.
    Width (Metric) 25.4 cm
    Temperature Range (Metric) -40°C to 80°C
    Temperature Range (English) -40°F to 175°F
    Displayed Text DANGER Corrosive
    Product Type Chemical and Hazardous Materials Sign
    Color Red on White
    Show More Show Less