missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Danger: Corrosive Signs

Identify equipment, rooms and materials that contain or are contaminated with hazardous agents.

Supplier:  Brady™ 70258

 View more versions of this product

Catalog No. 1704612



Only null left

Description

Description

  • Bright colors and bold text provide high visibility to critical messages
  • Sign reads DANGER CORROSIVE
  • Black/red on white
  • Designed to withstand harsh industrial environments
  • Available in different sizes and materials

B-401

  • Plastic — high-impact polystyrene
  • B-120
    • Premium fiberglass — fiberglass reinforced polyester
  • B-302
    • Self-sticking polyester — surface printed polyester, polyester overlaminate and sensitive acrylic

    Specifications

    Chemical Identifiers

    1 Ea.
    -40°F
    Chemical and Hazardous Materials Sign
    Surface Mount
    DANGER Corrosive

    Specifications

    ANSI Z535, OSHA 1910.145, ISO 9000, ASTM D 1593, ASTM D 638, ASTM D 638, ASTM D 646
    1 Ea.
    Premium Fiberglass
    Industrial, utility, commercial and institutional environment
    25.4 cm
    -40°F
    14 in.
    -40°C to 90°C
    DANGER Corrosive
    Black
    Fiberglass
    9000 psi
    Surface Mount
    10 in.
    193°F
    0.1 in.
    35.5 cm
    -40°F to 193°F
    Chemical and Hazardous Materials Sign
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Promotions

    Promotions