missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Danger: Confined Space -- Hazardous Atmosphere Signs
For identifying areas that require authorized or permit entry only
$20.40 - $20.40
Specifications
| Quantity | 1 Ea. |
|---|---|
| Mounting Type | Surface Mount |
| For Use With (Application) | Designed for use in industrial, utility, commercial and institutional environments |
| Min. Temperature (English) | -40°F |
| Displayed Text | Confined Space Hazardous Atmosphere Check Oxygen Level Before And During Entry |
| Catalog Number | Mfr. No. | Dimensions (L x W) | Material | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Dimensions (L x W) | Material | Price | Quantity | ||||
|
19-035-792
|
Brady™
BRD22430 |
14 x 10 in. | Plastic |
Each for $20.40
|
|
||||
Description
- Bright colors and bold text provide high visibility to critical messages
- Legend: CONFINED SPACE HAZARDOUS ATMOSPHERE CHECK OXYGEN LEVEL BEFORE AND DURING ENTRY
- Header: DANGER
- Black/red on white
- Designed to withstand harsh industrial environments
- Available in different sizes and materials
B-401
- Plastic — high-impact polystyrene
- Self-sticking polyester — surface printed polyester, polyester overlaminate and sensitive acrylic
Specifications
| 1 Ea. | |
| Designed for use in industrial, utility, commercial and institutional environments | |
| Confined Space Hazardous Atmosphere Check Oxygen Level Before And During Entry | |
| Red on White |
| Surface Mount | |
| -40°F | |
| Admittance Sign |