missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Danger: Compressed Gas Signs

Catalog No. 19035703
Change view
Click to view available options
Material:
Aluminum
Plastic
Polyester
Height (Metric):
17.78 cm
25.4 cm
Width (Metric):
25.4 cm
35.56 cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Height (Metric) Width (Metric)
19035703 Polyester 25.4 cm 35.56 cm
19102395 Plastic 17.78 cm 25.4 cm
19807021 Aluminum 25.4 cm 35.56 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19035703 Supplier Brady™ Supplier No. 84366

Identify equipment, rooms and materials that contain or are contaminated with hazardous agents

  • Bright colors and bold text provide high visibility for critical messages
  • Sign reads “DANGER COMPRESSED GAS”
  • Black/red on white
  • Designed to withstand harsh industrial environments
  • Available in different sizes and materials

B-401

  • Plastic — high-impact polystyrene
  • B-120
    • Premium fiberglass — fiberglass reinforced polyester
  • B-555
    • Aluminum — 0.1cm (0.040 in.) coil coated aluminum construction
  • B-302
    • Self-sticking polyester — surface printed polyester, polyester overlaminate and sensitive acrylic

    Specifications

    Certifications/Compliance ANSI Z535, OSHA 1910.145
    Material Polyester
    Quantity 1 Ea.
    Mounting Type Surface Mount
    For Use With (Application) Identification of fire alarms, extinguishers and emergency exits
    Height (English) 10 in.
    Height (Metric) 25.4 cm
    Max. Temperature (English) 230°F
    Min. Temperature (English) -40°F
    Thickness 0.006 to 0.01 in.
    Width (English) 14 in.
    Width (Metric) 35.56 cm
    Temperature Range (Metric) -40°C to 110°C
    Temperature Range (English) -40°F to 230°F
    Displayed Text DANGER Compressed Gas
    Product Type Chemical and Hazardous Materials Sign
    Color Red on White
    Show More Show Less