missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Danger: Chlorine Gas Signs

Catalog No. 19807020
Change view
Click to view available options
Dimensions (L x W):
10 x 14 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19807020 10 x 14 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19807020 Supplier Brady™ Supplier No. 40880

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Identify equipment, rooms and materials that contain or are contaminated with hazardous agents. Danger: Chlorine Gas signs are durable Non-Reflective Aluminum (B-555) panels with protected graphics for use in industrial, utility, commercial and institutional environments.

  • Bright colors and bold text provide high visibility to critical messages
  • Sign reads DANGER CHLORINE GAS
  • Black/red on white
  • Designed to withstand harsh industrial environments
  • Aluminum — 0.040 in. coil coated aluminum construction
  • Sign measures 10 in. H x 14 in. W

Specifications

Certifications/Compliance ANSI Z535, OSHA 1910.145
Material Aluminum
Quantity 1 Ea.
Adhesive Type Non-adhesive
Mounting Type Surface Mount
For Use With (Application) Industrial, utility, commercial and institutional environment
Height (English) 10 in.
Height (Metric) 25.4 cm
Max. Temperature (English) 230°F
Min. Temperature (English) -40°F
Thickness 0.04 in.
Width (English) 14 in.
Width (Metric) 35.56 cm
Temperature Range (Metric) -40°C to 110°C
Shape Rectangular
Temperature Range (English) -40°F to 230°F
Languages English
Dimensions (L x W) 10 x 14 in.
Displayed Text DANGER Chlorine Gas
Product Type Chemical and Hazardous Materials Sign
Color Red on White
Show More Show Less