missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Danger: Chemical Waste Storage Area… Signs

Catalog No. 19035582
Change view
Click to view available options
Material:
Plastic
Polyester
Height (English):
10 in.10 in.
10 in.
Width (English):
14 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Height (English) Width (English)
19035582 Plastic 10 in. 14 in.
19035584 Polyester 10 in.10 in. 14 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19035582 Supplier Brady™ Supplier No. BRD22080
Only null left

Identify equipment, rooms and materials that contain or are contaminated with hazardous agents

  • Bright colors and bold text provide high visibility to critical messages
  • Sign reads “DANGER CHEMICAL WASTE STORAGE AREA UNAUTHORIZED PERSONS KEEP OUT”
  • Black/red on white
  • Designed to withstand harsh industrial environments
  • Available in different sizes and materials

B-401

  • Plastic — high-impact polystyrene
  • B-302
    • Self-sticking polyester — surface printed polyester, polyester overlaminate and sensitive acrylic

    Specifications

    Certifications/Compliance ANSI Z535, OSHA 1910.145, ISO 9000, ASTM D 1593
    Material Plastic
    Quantity 1 Ea.
    For Use With (Application) Light duty industrial, utility, commercial, institutional environments
    Height (English) 10 in.
    Height (Metric) 25.4 cm
    Max. Temperature (English) 175°F
    Min. Temperature (English) -40°F
    Thickness 0.06 in.
    Width (English) 14 in.
    Width (Metric) 35.5 cm
    Temperature Range (Metric) -40°C to 80°C
    Temperature Range (English) -40°F to 175°F
    Displayed Text DANGER Chemical Waste Storage Area Unauthorized Persons Keep Out
    Product Type Admittance Sign
    Color Black
    Show More Show Less