missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Danger: Chemical Storage Signs

Catalog No. 19102111
Change view
Click to view available options
Material:
Fiberglass
Plastic
Polyester
Height (English):
7 in.
10 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Height (English)
19102111 Polyester 7 in.
1704610 Fiberglass 10 in.
19102110 Plastic 10 in.
19102109 Plastic 7 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19102111 Supplier Brady™ Supplier No. 84351

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Identify equipment, rooms and materials that contain or are contaminated with hazardous agents.

  • Bright colors and bold text provide high visibility to critical messages
  • Sign reads DANGER: CHEMICAL STORAGE
  • Black/red on white
  • Designed to withstand harsh industrial environments
  • Available in different sizes and materials

B-401

  • Plastic — high-impact polystyrene

B-120

  • Premium fiberglass — fiberglass reinforced polyester

B-302

  • Self-sticking polyester — surface printed polyester, polyester overlaminate and sensitive acrylic

Specifications

Certifications/Compliance ANSI Z535, OSHA 1910.145
Material Polyester
Quantity 1 Ea.
Mounting Type Surface Mount
For Use With (Application) Identification of fire alarms, extinguishers and emergency exits
Height (English) 7 in.
Height (Metric) 17.7 cm
Max. Temperature (English) 230°F
Min. Temperature (English) -40°F
Thickness 0.006 to 0.01 in.
Width (English) 10 in.
Width (Metric) 25.4 cm
Temperature Range (Metric) -40°C to 110°C
Temperature Range (English) -40°F to 230°F
Displayed Text DANGER Chemical Storage
Product Type Chemical and Hazardous Materials Sign
Color Red on White
Show More Show Less