missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Danger: Chemical Storage Signs
Identify equipment, rooms and materials that contain or are contaminated with hazardous agents.
Supplier: Brady™ 22312
Description
- Bright colors and bold text provide high visibility to critical messages
- Sign reads DANGER: CHEMICAL STORAGE
- Black/red on white
- Designed to withstand harsh industrial environments
- Available in different sizes and materials
B-401
- Plastic — high-impact polystyrene
B-120
- Premium fiberglass — fiberglass reinforced polyester
B-302
- Self-sticking polyester — surface printed polyester, polyester overlaminate and sensitive acrylic
Chemical Identifiers
| 1 Ea. | |
| -40°F | |
| Chemical and Hazardous Materials Sign |
| Surface Mount | |
| DANGER Chemical Storage |
Specifications
| ASTM D 1593 | |
| 1 Ea. | |
| Light duty industrial, utility, commercial, institutional environments | |
| 25.4 cm | |
| -40°F | |
| 14 in. | |
| -40°C to 80°C | |
| DANGER Chemical Storage | |
| Red on White |
| Plastic | |
| Surface Mount | |
| 10 in. | |
| 175°F | |
| 0.06 in. | |
| 35.5 cm | |
| -40°F to 175°F | |
| Chemical and Hazardous Materials Sign |