missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Danger: Chemical Storage Signs

Catalog No. 1704610
Change view
Click to view available options
Material:
Fiberglass
Plastic
Polyester
Height (English):
7 in.
10 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Height (English)
1704610 Fiberglass 10 in.
19102110 Plastic 10 in.
19102111 Polyester 7 in.
19102109 Plastic 7 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 1704610 Supplier Brady™ Supplier No. 70240

Identify equipment, rooms and materials that contain or are contaminated with hazardous agents.

  • Bright colors and bold text provide high visibility to critical messages
  • Sign reads DANGER: CHEMICAL STORAGE
  • Black/red on white
  • Designed to withstand harsh industrial environments
  • Available in different sizes and materials

B-401

  • Plastic — high-impact polystyrene

B-120

  • Premium fiberglass — fiberglass reinforced polyester

B-302

  • Self-sticking polyester — surface printed polyester, polyester overlaminate and sensitive acrylic

Specifications

Certifications/Compliance ANSI Z535, OSHA 1910.145, ISO 9000, ASTM D 1593, ASTM D 638, ASTM D 638, ASTM D 646
Material Fiberglass
Quantity 1 Ea.
Tensile Strength 9000 psi
Description Premium Fiberglass
Mounting Type Surface Mount
For Use With (Application) Industrial, utility, commercial and institutional environment
Height (English) 10 in.
Height (Metric) 25.4 cm
Max. Temperature (English) 193°F
Min. Temperature (English) -40°F
Thickness 0.1 in.
Width (English) 14 in.
Width (Metric) 35.5 cm
Temperature Range (Metric) -40°C to 90°C
Temperature Range (English) -40°F to 193°F
Displayed Text DANGER Chemical Storage
Product Type Chemical and Hazardous Materials Sign
Color Red on White
Show More Show Less