missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Compact Keyed-Different 1 in. Shackle Aluminum Padlocks

Catalog No. 19103517
Change view
Click to view available options
Color:
Black
Blue
Brown
Green
Orange
Purple
Red
Silver
Yellow
Quantity:
1 Ea.
6/Pk.
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Quantity
19103517 Green 6/Pk.
19103520 Black 6/Pk.
19103524 Red 1 Ea.
19103525 Blue 1 Ea.
19103526 Green 1 Ea.
19103527 Yellow 1 Ea.
19103528 Orange 1 Ea.
19103529 Black 1 Ea.
19103530 Brown 1 Ea.
19103531 Purple 1 Ea.
19103532 Silver 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 19103517 Supplier Brady™ Supplier No. 133263

Compact Keyed-Different 1 in. Shackle Aluminum Padlocks include two keys per lock.

  • Compact lock body measures 1-1/4 in. H x 1-1/4 in. W
  • Bright, anodized aluminum finish provides greater visibility and fade-resistance in all environoments
  • Reserved, paracentric keyway provides optimal security (prevents someone from getting a copy made at a local key shop)
  • Each padlock is keyed differently with keys precision-machined to exacting standards, so that the key for one lock will not open another in the set

Specifications

Certifications/Compliance OSHA 1910.147
Color Green
Key Type Keyed Different
Material Aluminum
Quantity 6/Pk.
Product Type Padlock
For Use With (Application) Outdoor; Industrial Manufacturing; Food and Beverage; Confined Spaces
For Use With (Equipment) Outdoor; Industrial Manufacturing; Food and Beverage; Confined Spaces
Width (English) Shackle 1 in.