Learn More
Brady™ Compact Keyed Alike 1 in. Shackle Aluminum Padlocks
Compact Keyed Alike 1 in. Shackle Aluminum Padlocks include two keys per lock.
Supplier: Brady™ 133284
Description
- Compact lock body measures 1-1/4 in. H x 1-1/4 in. W
- Bright, anodized aluminum finish provides greater visibility and fade-resistance in all environoments
- Reserved, paracentric keyway provides optimal security (prevents someone from getting a copy made at a local key shop)
- Every lock in this set can be opened with the same key, which is beneficial when multiple locks are assigned to a single employee
Specifications
| OSHA 1910.147 | |
| Keyed Alike | |
| 3/Pk. | |
| Outdoor; Industrial Manufacturing; Food and Beverage; Confined Spaces | |
| 1 in. |
| Black | |
| Aluminum | |
| Padlock | |
| Outdoor; Industrial Manufacturing; Food and Beverage; Confined Spaces |