missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Compact Keyed Alike 1 in. Shackle Aluminum Padlocks

Catalog No. 19103534
Change view
Click to view available options
Color:
Black
Blue
Brown
Green
Orange
Purple
Red
Silver
Yellow
Quantity:
3/Pk.
6/Pk.
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Quantity
19103534 Blue 3/Pk.
19103533 Red 3/Pk.
19103537 Orange 3/Pk.
19103542 Red 6/Pk.
19103543 Blue 6/Pk.
19103544 Green 6/Pk.
19103545 Yellow 6/Pk.
19103546 Orange 6/Pk.
19103547 Black 6/Pk.
19103548 Brown 6/Pk.
19103549 Purple 6/Pk.
19103550 Silver 6/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 19103534 Supplier Brady™ Supplier No. 133280
Only null left

Compact Keyed Alike 1 in. Shackle Aluminum Padlocks include two keys per lock.

  • Compact lock body measures 1-1/4 in. H x 1-1/4 in. W
  • Bright, anodized aluminum finish provides greater visibility and fade-resistance in all environoments
  • Reserved, paracentric keyway provides optimal security (prevents someone from getting a copy made at a local key shop)
  • Every lock in this set can be opened with the same key, which is beneficial when multiple locks are assigned to a single employee

Specifications

Certifications/Compliance OSHA 1910.147
Color Blue
Key Type Keyed Alike
Material Aluminum
Quantity 3/Pk.
Product Type Padlock
For Use With (Application) Outdoor; Industrial Manufacturing; Food and Beverage; Confined Spaces
For Use With (Equipment) Outdoor; Industrial Manufacturing; Food and Beverage; Confined Spaces
Width (English) Shackle 1 in.