Learn More
Brady™ Caution Hearing Protection Required in This Area Signs
Caution: Hearing Protection Required in This Area safety signs from Brady™ remind employees of proper procedures for sustaining a safe workplace. They are available in a variety of materials and sizes to best fit your application.
Supplier:  Brady™ 45071
 View more versions of this product
Catalog No. 19038055
Only null left
Description
Description
  • Plastic signs (B-401) are ideal for use in light duty industrial, utility, commercial and institutional environments.
  • Durable Non-Reflective Aluminum (B-555) is a rigid aluminum panel with protected graphics designed for use in industrial, utility, commercial and institutional environments.
Specifications
Chemical Identifiers
ASTM D 1593, ANSI Z535, OSHA 1910.145
Non-adhesive
-40°F
ANSI Z535 Safety Sign
1 Ea.
Surface Mount
CAUTION Hearing Protection Required In This Area
Specifications
ASTM D 1593, ANSI Z535, OSHA 1910.145
1 Ea.
Surface Mount
10 in.
-40°F
7 in.
-40°C to 80°C
10 x 7 in.
ANSI Z535 Safety Sign
Plastic
Non-adhesive
Light duty industrial, utility, commercial and institutional environments for both indoor and outdoor use
175°F
0.06 in.
177.8 mm
-40°F to 175°F
CAUTION Hearing Protection Required In This Area
Black on White
Product Suggestions
Product Suggestions