missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Caution: Hazardous Waste Storage Area… Signs

Catalog No. 19016659
Change view
Click to view available options
Material:
Fiberglass
Plastic
Height (English):
7 in.
10 in.
Width (English):
10 in.
14 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Height (English) Width (English)
19016659 Fiberglass 10 in. 14 in.
19035675 Plastic 7 in. 10 in.
19035676 Plastic 10 in. 14 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19016659 Supplier Brady™ Supplier No. 70374

Identify equipment, rooms and materials that contain or are contaminated with hazardous agents.

  • Bright colors and bold text provide high visibility to critical messages
  • Sign reads “CAUTION HAZARDOUS WASTE STORAGE AREA UNAUTHORIZED PERSONS KEEP OUT”
  • Black on yellow
  • Designed to withstand harsh industrial environments
  • Available in different sizes and materials

B-401

  • Plastic — high-impact polystyrene
  • B-120
    • Premium fiberglass — fiberglass reinforced polyester
  • B-302
    • Self-sticking polyester — surface printed polyester, polyester overlaminate and sensitive acrylic

    Specifications

    Certifications/Compliance ANSI Z535, OSHA 1910.145, ISO 9000, ASTM D 1593, ASTM D 638, ASTM D 638, ASTM D 646
    Material Fiberglass
    Quantity 1 Ea.
    Tensile Strength 9000 psi
    Description Premium Fiberglass
    Mounting Type Surface Mount
    For Use With (Application) Industrial, utility, commercial and institutional environment
    Height (English) 10 in.
    Height (Metric) 25.4 cm
    Max. Temperature (English) 193°F
    Min. Temperature (English) -40°F
    Thickness 0.1 in.
    Width (English) 14 in.
    Width (Metric) 35.5 cm
    Temperature Range (Metric) -40°C to 90°C
    Temperature Range (English) -40°F to 193°F
    Displayed Text CAUTION Hazardous Waste Storage Area Unauthorized Persons Keep Out
    Product Type Chemical and Hazardous Materials Sign
    Color Black on Yellow
    Show More Show Less