missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Caution: Hazardous Waste Materials Storage Only… Signs

Catalog No. 19035472
Change view
Click to view available options
Material:
Fiberglass
Plastic
Height (English):
7 in.
10 in.
14 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Height (English)
19035472 Plastic 10 in.
19035672 Plastic 7 in.
19036545 Fiberglass 14 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19035472 Supplier Brady™ Supplier No. BRD22708

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Identify equipment, rooms and materials that contain or are contaminated with hazardous agents

  • Bright colors and bold text provide high visibility to critical messages
  • Sign reads “CAUTION HAZARDOUS WASTE MATERIAL STORAGE ONLY NO SMOKING, OPEN FLAMES OR OPEN LIGHTS”
  • Black on yellow
  • Designed to withstand harsh industrial environments
  • Available in different sizes and materials

B-401

  • Plastic — high-impact polystyrene
  • B-120
    • Premium fiberglass — fiberglass reinforced polyester
  • B-302
    • Self-sticking polyester — surface printed polyester, polyester overlaminate and sensitive acrylic

    Specifications

    Certifications/Compliance ASTM D 1593,ANSI Z535, OSHA 1910.145
    Material Plastic
    Quantity 1 Ea.
    Mounting Type Surface Mount
    For Use With (Application) Light duty industrial, utility, commercial, institutional environments
    Height (English) 10 in.
    Height (Metric) 25.4 cm
    Max. Temperature (English) 175°F
    Min. Temperature (English) -40°F
    Thickness 0.06 in.
    Width (English) 14 in.
    Width (Metric) 35.5 cm
    Temperature Range (Metric) -40°C to 80°C
    Shape Rectangular
    Temperature Range (English) -40°F to 175°F
    Languages English
    Displayed Text CAUTION Hazardous Waste Material Storage Only No Smoking, Open Flames or Open Lights
    Product Type Chemical and Hazardous Materials Sign
    Color Black on Yellow
    Show More Show Less