Learn More
Brady™ Caution Eye Protection Signs
Caution: Eye Protection personal safety signs from Brady™ remind employees of proper procedures for sustaining a safe workplace. They are available in a variety of materials and sizes to best fit your application.
Supplier:  Brady™ 45144
 View more versions of this product
Catalog No. 19-038-042
Only null left
Explore available promotions
Description
Description
  • Self-Sticking Polyester (B-302) is an Indoor/Outdoor high performance pressure sensitive sign that is surface printed polyester with a polyester overlaminate.
  • Plastic signs (B-401) are ideal for use in light duty industrial, utility, commercial and institutional environments.
  • Durable Non-Reflective Aluminum (B-555) is a rigid aluminum panel with protected graphics designed for use in industrial, utility, commercial and institutional environments.
Specifications
Specifications
ANSI Z535, OSHA 1910.145, PSTC-33,
1 Ea.
Surface Mount
14 in.
-40°F
10 in.
-40°C to 110°C
14 x 10 in.
ANSI Z535 Safety Sign
Polyester
Permanent Acrylic
Identification of fire alarms, extinguishers and emergency exits for both indoor/outdoor applications and in harsh environments where excellent abrasion and chemical resistance is required beyond a surface printed label
230°F
0.006 to 0.01 in.
25.4 cm
-40°F to 230°F
CAUTION Eye Protection Required
Yellow on White
Product Suggestions
Product Suggestions